Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133241.2 | NP_573504.1 | 4, 5 | 373 | |
GenBank mRNA | AF449425.1 | - | 4, 5 | 827 | |
AK030396.1 | - | 4, 5 | 372 | ||
AK043757.1 | - | 4, 5 | 373 | ||
AK122190.1 | - | 4, 5 | 815 | ||
BC024719.1 | - | 4, 5 | 373 | 5363238 |