Gene Transcripts

Gene Symbol
COL17A1
Entrez Gene ID
1308
Gene Name
collagen type XVII alpha 1
Gene Aliases
BA16H23.2, BP180, BPA-2, BPAG2, ERED, LAD-1
Chromosome Location
Chr. 10: 104031288 - 104085880 on Build GRCh38
UniGene ID
Hs.117938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000494.3 NP_000485.3 5 476
GenBank mRNA AB900157.1 - 4 307
AK309940.1 - 5 422
BC004478.2 - 5 458
M91669.1 - 5 412

View Full Product Details