Gene Transcripts

Gene Symbol
Zcchc7
Entrez Gene ID
319885
Gene Name
zinc finger, CCHC domain containing 7
Gene Aliases
4930572I07Rik, D4Wsu132e
Chromosome Location
Chr. 4: 44756559 - 44932215 on Build GRCm38
UniGene ID
Mm.331576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138590.4 NP_613056.2 7, 8 1220
GenBank mRNA AK013175.1 - 1, 2 704
AK050430.1 - 1, 2 1912
BC054405.1 - 1, 2 423

View Full Product Details