gRNA Design Details

Target DNA Sequence
GAATCTGCACTGTGTGCCGG
PAM Sequence
TGG
Target locus
Chr.14: 74949380 - 74949402 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PGF
Entrez Gene ID
5228
Gene Name
placental growth factor
Gene Aliases
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760
Chromosome Location
Chr.14: 74941830 - 74955764 on GRCh38
UniGene ID
Hs.252820
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001207012.1 NP_001193941.1 3
NM_001293643.1 NP_001280572.1 3
NM_002632.5 NP_002623.2 3