gRNA Design Details
- Target DNA Sequence
- AACCTCGCCTCCTTTGTGAC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 159323130 - 159323152 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IL12B
- Entrez Gene ID
-
3593
- Gene Name
- interleukin 12B
- Gene Aliases
-
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
- Chromosome Location
-
Chr.5: 159314783 - 159330473 on GRCh38
- UniGene ID
- Hs.674
- Species
-
Human