gRNA Design Details
- Target DNA Sequence
- AACAGGCCAACCTCAACTCA
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 176729588 - 176729610 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.67/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- VEGFC
- Entrez Gene ID
-
7424
- Gene Name
- vascular endothelial growth factor C
- Gene Aliases
-
Flt4-L, LMPH1D, VRP
- Chromosome Location
-
Chr.4: 176683534 - 176792745 on GRCh38
- UniGene ID
- Hs.435215
- Species
-
Human