gRNA Design Details
- Target DNA Sequence
- ACTATGCGTCCCAGCAGCTT
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 15962978 - 15963000 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.83/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGFBP2
- Entrez Gene ID
-
83888
- Gene Name
- fibroblast growth factor binding protein 2
- Gene Aliases
-
HBP17RP, KSP37
- Chromosome Location
-
Chr.4: 15960240 - 15963236 on GRCh38
- UniGene ID
- Hs.98785
- Species
-
Human