gRNA Design Details
- Target DNA Sequence
- TGACCACTTCCCTGCAAGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 164621286 - 164621308 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.47/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GRB14
- Entrez Gene ID
-
2888
- Gene Name
- growth factor receptor bound protein 14
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 164492406 - 164621850 on GRCh38
- UniGene ID
- Hs.411881
- Species
-
Human