gRNA Design Details
- Target DNA Sequence
- GATACTTTCTAGGTCCGAGG
- PAM Sequence
- GGG
- Target locus
-
Chr.7: 23470048 - 23470070 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.63/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGF2BP3
- Entrez Gene ID
-
10643
- Gene Name
- insulin like growth factor 2 mRNA binding protein 3
- Gene Aliases
-
CT98, IMP-3, IMP3, KOC, KOC1, VICKZ3
- Chromosome Location
-
Chr.7: 23310209 - 23470674 on GRCh38
- UniGene ID
- Hs.700696
- Species
-
Human