gRNA Design Details

Target DNA Sequence
ATATCCCGCCTCATTTACAG
PAM Sequence
TGG
Target locus
Chr.7: 23418779 - 23418801 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.25/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGF2BP3
Entrez Gene ID
10643
Gene Name
insulin like growth factor 2 mRNA binding protein 3
Gene Aliases
CT98, IMP-3, IMP3, KOC, KOC1, VICKZ3
Chromosome Location
Chr.7: 23310209 - 23470674 on GRCh38
UniGene ID
Hs.700696
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_006547.2 NP_006538.2 3
XM_006715639.2 XP_006715702.1 2
XM_011515089.2 XP_011513391.1 3
XM_011515090.2 XP_011513392.1 2
XM_011515091.2 XP_011513393.1 3
XM_011515092.2 XP_011513394.1 3
XM_011515093.2 XP_011513395.1 2