gRNA Design Details

Target DNA Sequence
ATGGCTACGTTTCCTGTATG
PAM Sequence
TGG
Target locus
Chr.2: 111929275 - 111929297 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 90.57/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MERTK
Entrez Gene ID
10461
Gene Name
MER proto-oncogene, tyrosine kinase
Gene Aliases
MER, RP38, Tyro12, c-Eyk, c-mer
Chromosome Location
Chr.2: 111898479 - 112039946 on GRCh38
UniGene ID
Hs.306178
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_006343.2 NP_006334.2 2
XM_005263565.4 XP_005263622.1 2
XM_005263568.4 XP_005263625.1 2
XM_011510490.2 XP_011508792.1 2
XM_017003164.1 XP_016858653.1 2
XM_017003165.1 XP_016858654.1 2