gRNA Design Details

Target DNA Sequence
GGAGATCCGCTCTGTACACG
PAM Sequence
TGG
Target locus
Chr.19: 643242 - 643264 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 94.55/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF22
Entrez Gene ID
27006
Gene Name
fibroblast growth factor 22
Gene Aliases
-
Chromosome Location
Chr.19: 639895 - 643703 on GRCh38
UniGene ID
Hs.248087
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001300812.1 NP_001287741.1 2
NM_020637.1 NP_065688.1 2