gRNA Design Details
- Target DNA Sequence
- GTGAAGCGTGTTCTCTACAT
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 45913463 - 45913485 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
93.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP3
- Entrez Gene ID
-
3486
- Gene Name
- insulin like growth factor binding protein 3
- Gene Aliases
-
BP-53, IBP3
- Chromosome Location
-
Chr.7: 45912245 - 45921272 on GRCh38
- UniGene ID
- Hs.450230
- Species
-
Human