gRNA Design Details
- Target DNA Sequence
- AACATGTGGTGAGCATTCCA
- PAM Sequence
- CGG
- Target locus
-
Chr.7: 45913533 - 45913555 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
93.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP3
- Entrez Gene ID
-
3486
- Gene Name
- insulin like growth factor binding protein 3
- Gene Aliases
-
BP-53, IBP3
- Chromosome Location
-
Chr.7: 45912245 - 45921272 on GRCh38
- UniGene ID
- Hs.450230
- Species
-
Human