gRNA Design Details
- Target DNA Sequence
- GACCCATTTGGGCTTGTCAC
- PAM Sequence
- CGG
- Target locus
-
Chr.11: 69699299 - 69699321 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
93.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF19
- Entrez Gene ID
-
9965
- Gene Name
- fibroblast growth factor 19
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 69698238 - 69704338 on GRCh38
- UniGene ID
- Hs.249200
- Species
-
Human