gRNA Design Details
- Target DNA Sequence
- GGTACACATTGTAGCCATCT
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 69699510 - 69699532 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.51/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF19
- Entrez Gene ID
-
9965
- Gene Name
- fibroblast growth factor 19
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 69698238 - 69704338 on GRCh38
- UniGene ID
- Hs.249200
- Species
-
Human