gRNA Design Details
- Target DNA Sequence
- GAGCACAAGATCTCCTACAG
- PAM Sequence
- CGG
- Target locus
-
Chr.17: 48997815 - 48997837 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
68.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGF2BP1
- Entrez Gene ID
-
10642
- Gene Name
- insulin like growth factor 2 mRNA binding protein 1
- Gene Aliases
-
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1
- Chromosome Location
-
Chr.17: 48997412 - 49056145 on GRCh38
- UniGene ID
- Hs.144936
- Species
-
Human