gRNA Design Details
- Target DNA Sequence
- CACATAAACACCGGCATCCT
- PAM Sequence
- CGG
- Target locus
-
Chr.5: 108703478 - 108703500 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgfrl1
- Entrez Gene ID
-
116701
- Gene Name
- fibroblast growth factor receptor-like 1
- Gene Aliases
-
FGFR5, FGFR5beta, FGFR5gamma
- Chromosome Location
-
Chr.5: 108694229 - 108706950 on GRCm38
- UniGene ID
- Mm.35691
- Species
-
Mouse