gRNA Design Details

Target DNA Sequence
CACATAAACACCGGCATCCT
PAM Sequence
CGG
Target locus
Chr.5: 108703478 - 108703500 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 83.19/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Fgfrl1
Entrez Gene ID
116701
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FGFR5beta, FGFR5gamma
Chromosome Location
Chr.5: 108694229 - 108706950 on GRCm38
UniGene ID
Mm.35691
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_054071.2 NP_473412.1 3