gRNA Design Details

Target DNA Sequence
CGTTGGGGAAGATCTCAAGG
PAM Sequence
TGG
Application
Gene Knock-Out
Score: 87.49/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Fgf16
Entrez Gene ID
80903
Gene Name
fibroblast growth factor 16
Gene Aliases
-
Chromosome Location
Chr.X: 105764477 - 105776532 on GRCm38
UniGene ID
Mm.154768
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_030614.2 NP_085117.2 1