gRNA Design Details
- Target DNA Sequence
- CGTTGGGGAAGATCTCAAGG
- PAM Sequence
- TGG
- Application
- Gene Knock-Out
Score:
87.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf16
- Entrez Gene ID
-
80903
- Gene Name
- fibroblast growth factor 16
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 105764477 - 105776532 on GRCm38
- UniGene ID
- Mm.154768
- Species
-
Mouse