gRNA Design Details
- Target DNA Sequence
- GGTAATAGGCGGCCAGCGTA
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 24695357 - 24695379 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gfer
- Entrez Gene ID
-
11692
- Gene Name
- growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration)
- Gene Aliases
-
Alr, ERV1
- Chromosome Location
-
Chr.17: 24693189 - 24696227 on GRCm38
- UniGene ID
- Mm.28124
- Species
-
Mouse