gRNA Design Details
- Target DNA Sequence
- TACTTCCTCCTCAAGAACGA
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 112662257 - 112662279 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Akt1
- Entrez Gene ID
-
11651
- Gene Name
- thymoma viral proto-oncogene 1
- Gene Aliases
-
Akt, PKB, PKB/Akt, PKBalpha, Rac
- Chromosome Location
-
Chr.12: 112653821 - 112674802 on GRCm38
- UniGene ID
- Mm.6645
- Species
-
Mouse