gRNA Design Details
- Target DNA Sequence
- GCACGGCATTGGACATGTAG
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 18741083 - 18741105 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Angpt2
- Entrez Gene ID
-
11601
- Gene Name
- angiopoietin 2
- Gene Aliases
-
Agpt2, Ang-2, Ang2
- Chromosome Location
-
Chr.8: 18690263 - 18741562 on GRCm38
- UniGene ID
- Mm.439874
- Species
-
Mouse