gRNA Design Details
- Target DNA Sequence
- GTGTTACTGAGCATCGGTGA
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 4249464 - 4249486 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.24/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfbr3l
- Entrez Gene ID
-
100044509
- Gene Name
- transforming growth factor, beta receptor III-like
- Gene Aliases
-
Gm14378, OTTMUSG00000016477
- Chromosome Location
-
Chr.8: 4248214 - 4251423 on GRCm38
- UniGene ID
- Mm.439193
- Species
-
Mouse