gRNA Design Details

Target DNA Sequence
GTGTTACTGAGCATCGGTGA
PAM Sequence
AGG
Target locus
Chr.8: 4249464 - 4249486 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 83.24/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Tgfbr3l
Entrez Gene ID
100044509
Gene Name
transforming growth factor, beta receptor III-like
Gene Aliases
Gm14378, OTTMUSG00000016477
Chromosome Location
Chr.8: 4248214 - 4251423 on GRCm38
UniGene ID
Mm.439193
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001195258.1 NP_001182187.1 3