gRNA Design Details
- Target DNA Sequence
- GGCTCGTATACTGCCTCACT
- PAM Sequence
- CGG
- Target locus
-
Chr.5: 145136949 - 145136971 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pdap1
- Entrez Gene ID
-
231887
- Gene Name
- PDGFA associated protein 1
- Gene Aliases
-
HASPP28, PAP, PAP1
- Chromosome Location
-
Chr.5: 145128770 - 145140089 on GRCm38
- UniGene ID
- Mm.188851
- Species
-
Mouse