gRNA Design Details
- Target DNA Sequence
- AGCGCGGACGGCAAGATATA
- PAM Sequence
- CGG
- Target locus
-
Chr.7: 144897193 - 144897215 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf15
- Entrez Gene ID
-
14170
- Gene Name
- fibroblast growth factor 15
- Gene Aliases
-
FGF19
- Chromosome Location
-
Chr.7: 144896532 - 144900953 on GRCm38
- UniGene ID
- Mm.3904
- Species
-
Mouse