gRNA Design Details
- Target DNA Sequence
- CCCGTGCATCAATGTACGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 99041762 - 99041784 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igfbp4
- Entrez Gene ID
-
16010
- Gene Name
- insulin-like growth factor binding protein 4
- Gene Aliases
-
AI875747, Deb2, IGFBP-4
- Chromosome Location
-
Chr.11: 99041260 - 99052646 on GRCm38
- UniGene ID
- Mm.233799
- Species
-
Mouse