gRNA Design Details

Target DNA Sequence
TCCGCCAGAAAGTAGTGTGT
PAM Sequence
GGG
Target locus
Chr.8: 4354099 - 4354121 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 89.46/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Ccl25
Entrez Gene ID
20300
Gene Name
chemokine (C-C motif) ligand 25
Gene Aliases
A130072A22Rik, AI852536, CKb15, Scya25, TECK
Chromosome Location
Chr.8: 4325159 - 4360020 on GRCm38
UniGene ID
Mm.7275
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_009138.3 NP_033164.1 4
NR_033527.1 4