gRNA Design Details
- Target DNA Sequence
- GGTGATAATTCCTAGCATGC
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 4353938 - 4353960 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.11/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ccl25
- Entrez Gene ID
-
20300
- Gene Name
- chemokine (C-C motif) ligand 25
- Gene Aliases
-
A130072A22Rik, AI852536, CKb15, Scya25, TECK
- Chromosome Location
-
Chr.8: 4325159 - 4360020 on GRCm38
- UniGene ID
- Mm.7275
- Species
-
Mouse