gRNA Design Details
- Target DNA Sequence
- TTGAAACAGAGGGTCACTCG
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 27064307 - 27064329 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prl
- Entrez Gene ID
-
19109
- Gene Name
- prolactin
- Gene Aliases
-
AV290867, Prl1a1
- Chromosome Location
-
Chr.13: 27057570 - 27065204 on GRCm38
- UniGene ID
- Mm.1270
- Species
-
Mouse