gRNA Design Details

Target DNA Sequence
GGGTGTTATCAGCCAGTTCT
PAM Sequence
AGG
Target locus
Chr.7: 25787352 - 25787374 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 85.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Axl
Entrez Gene ID
26362
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
AI323647, Ark, Tyro7, Ufo
Chromosome Location
Chr.7: 25756500 - 25788733 on GRCm38
UniGene ID
Mm.4128
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001190974.1 NP_001177903.1 2
NM_001190975.1 NP_001177904.1 2
NM_009465.4 NP_033491.2 2