gRNA Design Details
- Target DNA Sequence
- GGGTGTTATCAGCCAGTTCT
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 25787352 - 25787374 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Axl
- Entrez Gene ID
-
26362
- Gene Name
- AXL receptor tyrosine kinase
- Gene Aliases
-
AI323647, Ark, Tyro7, Ufo
- Chromosome Location
-
Chr.7: 25756500 - 25788733 on GRCm38
- UniGene ID
- Mm.4128
- Species
-
Mouse