gRNA Design Details
- Target DNA Sequence
- GATCTGTCCCTTCCAAGTCC
- PAM Sequence
- TGG
- Target locus
-
Chr.18: 36515102 - 36515124 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Hbegf
- Entrez Gene ID
-
15200
- Gene Name
- heparin-binding EGF-like growth factor
- Gene Aliases
-
AW047313, Dtr, Dts, Hegfl
- Chromosome Location
-
Chr.18: 36504927 - 36515805 on GRCm38
- UniGene ID
- Mm.289681
- Species
-
Mouse