gRNA Design Details
- Target DNA Sequence
- GCAGTCCGTTGCTAACCAGT
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 109312314 - 109312336 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Eps15
- Entrez Gene ID
-
13858
- Gene Name
- epidermal growth factor receptor pathway substrate 15
- Gene Aliases
-
2410112D09Rik
- Chromosome Location
-
Chr.4: 109280256 - 109387817 on GRCm38
- UniGene ID
- Mm.318250
- Species
-
Mouse