gRNA Design Details
- Target DNA Sequence
- AAAGTGCGGCATGCTCAGAG
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 60943223 - 60943245 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Map2k3
- Entrez Gene ID
-
26397
- Gene Name
- mitogen-activated protein kinase kinase 3
- Gene Aliases
-
AW212142, MEK3, MKK3, Prkmk3, mMKK3b
- Chromosome Location
-
Chr.11: 60932057 - 60952803 on GRCm38
- UniGene ID
- Mm.18494
- Species
-
Mouse