gRNA Design Details

Target DNA Sequence
CATCCTGGAGATTACTGCGG
PAM Sequence
TGG
Target locus
Chr.7: 144840672 - 144840694 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 96.98/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Fgf3
Entrez Gene ID
14174
Gene Name
fibroblast growth factor 3
Gene Aliases
Fgf-3, Int-2, Int-P
Chromosome Location
Chr.7: 144837521 - 144844447 on GRCm38
UniGene ID
Mm.4947
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_008007.2 NP_032033.2 2