gRNA Design Details
- Target DNA Sequence
- ACAAGAGGGGCAAGCTGATT
- PAM Sequence
- GGG
- Target locus
-
Chr.14: 70638470 - 70638492 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf17
- Entrez Gene ID
-
14171
- Gene Name
- fibroblast growth factor 17
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 70636203 - 70643014 on GRCm38
- UniGene ID
- Mm.12814
- Species
-
Mouse