gRNA Design Details
- Target DNA Sequence
- TAACCAGTACGTGAGGGACC
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 22046140 - 22046162 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
71.11/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF17
- Entrez Gene ID
-
8822
- Gene Name
- fibroblast growth factor 17
- Gene Aliases
-
FGF-13, FGF-17, HH20
- Chromosome Location
-
Chr.8: 22039708 - 22048809 on GRCh38
- UniGene ID
- Hs.248192
- Species
-
Human