gRNA Design Details
- Target DNA Sequence
- GCGCTCATCAGGACAAACAC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 107266211 - 107266233 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.1/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfbr3
- Entrez Gene ID
-
21814
- Gene Name
- transforming growth factor, beta receptor III
- Gene Aliases
-
1110036H20Rik, AU015626, AW215636, TBRIII
- Chromosome Location
-
Chr.5: 107106570 - 107289595 on GRCm38
- UniGene ID
- Mm.200775
- Species
-
Mouse