gRNA Design Details
- Target DNA Sequence
- GACATTGACGGAGAAGTAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 25555748 - 25555770 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgfr1
- Entrez Gene ID
-
14182
- Gene Name
- fibroblast growth factor receptor 1
- Gene Aliases
-
AW208770, Eask, FLG, Fgfr-1, Flt-2, Hspy, MFR, bFGF-R-1, c-fgr
- Chromosome Location
-
Chr.8: 25513623 - 25575718 on GRCm38
- UniGene ID
- Mm.265716
- Species
-
Mouse