gRNA Design Details
- Target DNA Sequence
- CTACTGCAAGGTGCTACTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 49625344 - 49625366 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Flt4
- Entrez Gene ID
-
14257
- Gene Name
- FMS-like tyrosine kinase 4
- Gene Aliases
-
AI323512, Chy, Flt-4, VEGFR-3, VEGFR3
- Chromosome Location
-
Chr.11: 49609679 - 49652739 on GRCm38
- UniGene ID
- Mm.3291
- Species
-
Mouse