gRNA Design Details
- Target DNA Sequence
- AAAGCGGACGATTCTGAAGT
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 186649849 - 186649871 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfb2
- Entrez Gene ID
-
21808
- Gene Name
- transforming growth factor, beta 2
- Gene Aliases
-
BB105277, Tgf-beta2, Tgfb-2
- Chromosome Location
-
Chr.1: 186622787 - 186709033 on GRCm38
- UniGene ID
- Mm.18213
- Species
-
Mouse