gRNA Design Details
- Target DNA Sequence
- GAGATCTCCACGGTAGAACG
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 127017002 - 127017024 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf6
- Entrez Gene ID
-
14177
- Gene Name
- fibroblast growth factor 6
- Gene Aliases
-
Fgf-6, HSTF-2
- Chromosome Location
-
Chr.6: 127015542 - 127028182 on GRCm38
- UniGene ID
- Mm.3403
- Species
-
Mouse