gRNA Design Details
- Target DNA Sequence
- GTTGCAGCTCTCCAGCGACA
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 81037523 - 81037545 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pdgfc
- Entrez Gene ID
-
54635
- Gene Name
- platelet-derived growth factor, C polypeptide
- Gene Aliases
-
1110064L01Rik, AI647969, PDGF-C
- Chromosome Location
-
Chr.3: 81035885 - 81214040 on GRCm38
- UniGene ID
- Mm.331089
- Species
-
Mouse