gRNA Design Details

Target DNA Sequence
GGTCTTTCCGGTGAGAGGTC
PAM Sequence
TGG
Target locus
Chr.17: 46018795 - 46018817 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 91.65/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Vegfa
Entrez Gene ID
22339
Gene Name
vascular endothelial growth factor A
Gene Aliases
Vegf, Vpf
Chromosome Location
Chr.17: 46016993 - 46032377 on GRCm38
UniGene ID
Mm.282184
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001025250.3 NP_001020421.2 8
NM_001025257.3 NP_001020428.2 6
NM_001110266.1 NP_001103736.1 5
NM_001110267.1 NP_001103737.1 4
NM_001110268.1 NP_001103738.1 3
NM_001287056.1 NP_001273985.1 8
NM_001287057.1 NP_001273986.1 7
NM_001287058.1 NP_001273987.1 6
NM_009505.4 NP_033531.3 7