gRNA Design Details

Target DNA Sequence
TCGGACTTGGTGTTGCCCAG
PAM Sequence
TGG
Target locus
Chr.19: 6986296 - 6986318 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 84.14/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Vegfb
Entrez Gene ID
22340
Gene Name
vascular endothelial growth factor B
Gene Aliases
VEGF-B, Vrf
Chromosome Location
Chr.19: 6982472 - 6987651 on GRCm38
UniGene ID
Mm.15607
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001185164.1 NP_001172093.1 3
NM_011697.3 NP_035827.1 3