gRNA Design Details
- Target DNA Sequence
- ACACATTCCAGGCCATCGTC
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 6986321 - 6986343 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.51/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Vegfb
- Entrez Gene ID
-
22340
- Gene Name
- vascular endothelial growth factor B
- Gene Aliases
-
VEGF-B, Vrf
- Chromosome Location
-
Chr.19: 6982472 - 6987651 on GRCm38
- UniGene ID
- Mm.15607
- Species
-
Mouse