gRNA Design Details

Target DNA Sequence
GTAGCCTGATCCGACAGAAG
PAM Sequence
CGG
Target locus
Chr.11: 69801587 - 69801609 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 93.73/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Fgf11
Entrez Gene ID
14166
Gene Name
fibroblast growth factor 11
Gene Aliases
Fgf-11, Fhf-3, Fhf3
Chromosome Location
Chr.11: 69796068 - 69802945 on GRCm38
UniGene ID
Mm.269011
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001291104.1 NP_001278033.1 1
NM_010198.2 NP_034328.1 1